Potassium dichromate solution_standardised 0.25 N aqueous solution

Supplier: Thermo Fisher Scientific
Danger

35587.K2 35587.AP
35587.K2EA 54.6 EUR
35587.K2 35587.AP
Potassium dichromate solution_standardised 0.25 N aqueous solution
Potassium dichromate

CAS No.: 7778-50-9 Molecular Formula: Cr2K2O7 Formula Weight: 294.18 Physical Form: Liquid Appearance: Orange Odour: Odourless UN No.: 3287 MDL No.: MFCD00011367

Formula: K₂Cr₂O₇
MW: 294.18 g/mol
Boiling Pt: 500
Melting Pt: 398
Storage Temperature: Ambient
MDL Number: MFCD00011367
CAS Number: 7778-50-9
EINECS: 231-906-6
UN: 3287
ADR: 6.1 III
Merck Index: 12,07790

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR