Titanium 99,6 Metals basis, foil, annealed, Thickness: 0.05 mm (0.002 in)

Supplier: Thermo Fisher Scientific
Warning

35811.FF 35811.FI 35811.GH
35811.FFEA 70 EUR
35811.FF 35811.FI 35811.GH
Titanium 99,6 Metals basis, foil, annealed, Thickness: 0.05 mm (0.002 in)
Titanium

Purity: 99.6% CAS No.: 7440-32-6 Physical Form: ≈0.14g/25x25mm, Temper: soft Odour: Odourless MDL No.: MFCD00011264

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR