Acrylamide 99,9

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Prop-2-enamide

36545.14 36545.22
36545.14EA 60.7 EUR
36545.14 36545.22
Acrylamide 99,9
Acrylamide (monomer)

Purity: 99.9% CAS No.: 79-06-1 Molecular Formula: CH2=CHCONH2 Formula Weight: 71.08 Flash Point: 138°(280°F) Physical Form: Crystalline Density: 1.322 Sensitivity: Air and Light Sensitive Odour: Odourless Melting Point: 82-85° Boiling Point: 125°/25mm Beilstein Reference: 605349 Merck Reference: 14,129 UN No.: 2074 MDL No.: MFCD00008032

Formula: C₃H₅NO
MW: 71.08 g/mol
Boiling Pt: 125 °C (25 mmHg)
Melting Pt: 82…85 °C
Density: 1.322
Flash Pt: 138 °C (280 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008032
CAS Number: 79-06-1
EINECS: 201-173-7
UN: 2074
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7687b7f17a8cdf31c559a33fd5d4021a2e50a40a&vid=fecebe0c34b037788df119c11bfc9637d78cda0d&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR