Potassium-tert-butoxide 1 M THF, AcroSeal®, pure

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Potassium-2-methylpropan-2-olate, Potassium tert-butylate, Potassium tert-butanolate, Potassium t-butoxide

371221000 371220010 371220025
371221000.EA 207 EUR
371221000. 371220010. 371220025.
Potassium-tert-butoxide 1 M THF, AcroSeal®, pure
Potassium-tert-butoxide
Formula: C₄H₉KO
MW: 112.21 g/mol
Density: 0.902 g/cm³
Flash Pt: –19 °C
Storage Temperature: Ambient
MDL Number: MFCD00012162
CAS Number: 865-47-4
EINECS: 212-740-3
UN: 2920
ADR: 8 I

Order Now

Specification Test Results

Appearance Colorless to yellow to light orange Solution (May develop some turbidity or precipitate)
Strength 0.95 to 1.10 M

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR