4'-Hydroxybiphenyl-4-carboxylic acid 99%, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY

Synonyms: 4'-Hydroxy-4-biphenylcarboxylic acid, 4'-Hydroxy-[1,1'-biphenyl]-4-carboxylic acid

MQ100

371335-5G
371335-5GEA 176 EUR
371335-5G
4'-Hydroxybiphenyl-4-carboxylic acid 99%, Sigma-Aldrich®
4'-Hydroxybiphenyl-4-carboxylic acid
Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
Melting Pt: 295 °C (dec., lit.)
Storage Temperature: Room Temperature
MDL Number: MFCD00059078
CAS Number: 58574-03-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aecba806fa2dbf3d64011087bff6b5f3eae1aff4&vid=48fda0f30834b0816d2975033e2dab6d1bb767f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (colour) White to Light Brown
Appearance (form) Powder or Crystals or Powder with Chunk(s) or Crystals with Chunk(s)
Titration (T) NaOH 0.1 m 98.5 - 101.5%
Purity (HPLC area%) ≥ 98.5%
Infrared spectrum Conforms to structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR