Sodium periodate 99,8-100,3 Iodometry, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Sodium metaperiodate

M.O.C. Category N

3756-01EA 1235.87 EUR
3756-01 3756-04
Sodium periodate 99,8-100,3 Iodometry, BAKER ANALYZED® ACS, J.T.Baker®
Sodium periodate

  • Lot analysis on label

Formula: NaIO₄
MW: 213.89 g/mol
Boiling Pt: 300
Melting Pt: 175
Density: 3.87 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003534
CAS Number: 7790-28-5
EINECS: 232-197-6
UN: 1479
ADR: 5.1 II
Merck Index: 13,08714

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements 99.8 - 100.3%
For Laboratory,Research,or Manufacturing Use ≤0.02%
Assay (NaIO₄) (by iodometry) 99.8 - 100.3 %
Other Halogens (as Cl) ≤0.02 %
Trace Impurities - Manganese (Mn) ≤3.000 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR