Strontium titanate substrate 10x10x0.5 mm, polished one side 100 orientation

Supplier: Thermo Fisher Scientific

38506.KT 38506.KU
38506.KTEA 168 EUR
38506.KT 38506.KU
Strontium titanate substrate 10x10x0.5 mm, polished one side 100 orientation
Strontium titanate

CAS No.: 12060-59-2 Molecular Formula: SrTiO3 Odour: Odourless MDL No.: MFCD00049554

Formula: O₃SrTi
MW: 183.5 g/mol
Melting Pt: 2080
Density: 5.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049554
CAS Number: 12060-59-2
EINECS: 235-044-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR