Cooled Incubators, KB Series
Supplier: Binder
9020-0471 9020-0472 9020-0473 9020-0474 9020-0475 9020-0476
390-1741EA 7400 EUR
390-1741 390-1742 390-1743 390-1744 390-1745 390-1746
Cooled Incubators, KB Series
Incubators Cooled Incubators
The powerful KB Series of cooling incubators for microorganisms are able to incubate with exceptional precision even when fully loaded. Units have a stainless-steel inner chamber, feature proven APT.line™ preheating chamber technology, have a temperature range of 0 to +70 °C, hot-air disinfection routine, and can be programmed for weekend operation with ease. This KB unit consumes as much as 40% less energy than the previous model.
  • Energy-efficient cooling with modern inverter cooling unit
  • Cooling with climate-neutral R-600a refrigerant (GWP is 3)
  • Door
More Product Information
Order Now
SPECIFICATIONS
Model KB 65 KB 130 KB 260 KB 470 KB 720 KB 1060
Environmentally Preferable
Temperature fluctuation (time) ±0.2 K (at 37 °C) ±0.1 K (at 37 °C) ±0.1 K (at 37 °C) ±0.1 K (at 37 °C) ±0.1 K (at 37 °C) ±0.1 K (at 37 °C)
Temperature variation (spatial) ±0.2 K (at 37 °C) ±0.2 K (at 37 °C) ±0.2 K (at 37 °C) ±0.2 K (at 37 °C) ±0.2 K (at 37 °C) ±0.2 K (at 37 °C)
Temperature range 0...70 °C 0...70 °C 0...70 °C 0...70 °C 0...70 °C 0...70 °C
Capacity 63 L 127 L 262 L 472 L 727 L 1070 L
Convection type Compressor cooling and heating elements Compressor cooling and heating elements Compressor cooling and heating elements Compressor cooling and heating elements Compressor cooling and heating elements Compressor cooling and heating elements
Nominal power consumption 0.9 kW 1.2 kW 1.3 kW 1.3 kW 1.5 kW 1.5 kW
Weight 95 kg 124 kg 174 kg 212 kg 285 kg 315 kg
Shelves supplied (max.) 2 (4) 2 (6) 2 (9) 2 (16) 2 (16) 2 (16)
Max. load per shelf 25 kg 25 kg 30 kg 40 kg 40 kg 40 kg
Electrical 208 - 240V, 50/60 Hz, 16 Amp 208 - 240V, 50/60 Hz, 16 Amp 208 - 240V, 50/60 Hz, 16 Amp 208 - 240V, 50/60 Hz, 16 Amp 208 - 240V, 50/60 Hz, 16 Amp 208 - 240V, 50/60 Hz, 16 Amp
Doors 1 inner, 1 outer 1 inner, 1 outer 1 inner, 1 outer 1 inner, 1 outer 2 inner, 2 outer 2 inner, 2 outer