Cell culture dishes, glass bottom, CELLview™
Supplier: Greiner Bio-One
627860_40 627861_40 627870_40 627871_40 627965_40 627975_40
391-0252EA 438 EUR
391-0252 391-0253 391-0254 391-0255 391-0256 391-0257
Cell culture dishes, glass bottom, CELLview™
Culture Dishes Cell Culture Dishes
PS with integrated glass bottom, sterile. CELLview™ glass bottom dishes combine the convenience of a standard 35 mm disposable plastic cell culture dish with the optical quality of glass, providing superior high resolution microscopic images of in vitro cultures. The CELLview™ dish is also available as a subdivided version with four individual compartments facilitating simultaneous multiplex analysis of different cell lines, various stimulations or diverse transfections, minimising the amount of required cells and reagents per individual assay.
  • High transparent achromatic borosilicate
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001 (241002); ISO 50001 (241002)