Petri dishes, Sterilin®
Supplier: Sterilin
124-17 122-17 123 121V PF55 PF55V
391-2022EA 581 EUR
391-2022 391-2020 391-2021 391-2019 391-2041 391-2042
Petri dishes, Sterilin®
Culture Dishes Petri Dishes
Ideal for use when savings in media or incubator space are required.
  • Aseptically manufactured under cleanroom conditions (Class 7 ISO 14644) to exclude microbiological contamination
  • 55 mm dish accommodates 47 mm membrane filters making it suitable for water testing
  • 50 mm deep form dish is over 20 mm deep and designed for use with liquid media
Dimensions shown are for outer Ø base and outer height (mm).
Save incubator space with Thermo Scientific™ Sterilin™ 30 to 140 mm Petri Dishes. These small dishes are suitable for most applications. NotMore Product Information