Contact plate, with grid, ContactClik™
Supplier: PHOENIX BIOMEDICAL
116
391-3561EA 182 EUR
391-3561
Contact plate, with grid, ContactClik™
Culture Dishes Petri Dishes
Used for testing bioburden counts on working surfaces.
  • ContactClik™ design allows easy locking and reopening, no orientation required
  • Domed surface and media gripping tabs help the media to stick to the dish, reducing drop-off
  • Special double ring around the outside of the dish helps prevent fingers from touching the media, thus reducing the possibilities of contamination
  • Pre-treated all around the sides of the base and ready for printing a barcode, lot number or any other information as required