Storage vials, Nunc™
Supplier: Thermo Fisher Scientific
264262 264300
391-8348EA 0 EUR
391-8348 391-7589
Storage vials, Nunc™
Vials
High purity PPCO (PP copolymer) vial, PE screw cap. Excellent for centrifugation and storage of PCR reagents, restriction enzymes and other diagnostic and biochemical reagents or samples. Available in 0,5 and 2,0 ml sizes with frosted writing area; furthermore 2,0 ml size have graduations at 0,5 and 1,0 ml.
  • Leakproof screw closure even without an O-ring
  • One full turn removes the closure, giving easy access to contents
Can be centrifuged to a max. of 13000×g.
Can be used from −40 to +95 °C.
Can be used from -40 to +95 °C.