Iron ≥99.5% (metals basis), foil, Thickness: 0.127 mm (0.005 in)

Supplier: Thermo Fisher Scientific

39707.FF 39707.FQ
39707.FFEA 50.2 EUR
39707.FF 39707.FQ
Iron ≥99.5% (metals basis), foil, Thickness: 0.127 mm (0.005 in)
Iron

Purity: 99.5% CAS No.: 7439-89-6 Physical Form: ≈0.62g/25x25mm MDL No.: MFCD00010999

Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000
Melting Pt: 1535
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR