Aminoallyl-dUTP (AA-dUTP) sodium salt 10 mM in TE buffer

Supplier: Biotium

40020
40020.EA 265 EUR
40020.
Aminoallyl-dUTP (AA-dUTP) sodium salt 10 mM in TE buffer
Aminoallyl-dUTP (AA-dUTP) sodium salt

5-Aminoallyl-dUTP (AA-dUTP) can be enzymatically incorporated into DNA. The resulting amine-containing DNA can be subsequently labeled with a fluorescent dye, biotin or other haptens via conventional peptide coupling method.


This two-step method for labeling nucleic acids is considerably more economical than the one-step method using a prelabeled dUTP. Available in lyophilized solid form (1 mg), or as a 10 mM solution in TE Buffer (100 uL).

Formula: C₁₂H₁₇N₃Na₃O₁₄P₃
MW: 589.17 g/mol
Storage Temperature: Freezer
CAS Number: 116840-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR