BactoSpore™ stain_nucleic acid 500X in DMSO
Supplier: Biotium
Synonyms: Bacterial Stains
40120-T 40120
40120-TEA 285 EUR
40120-T 40120.
BactoSpore™ stain_nucleic acid 500X in DMSO
BactoSpore™
BactoSpore™ 488/540 nucleic acid stain is a yellow fluorescent nucleic acid dye that is detectable in both the FITC channel and the PE channel for flow cytometry.
  • Bacterial Stain
  • For nucleic acid staining in the FITC or PE channel
  • Optimised for staining endospores, unlike other bacterial detection reagents
  • Validated for microscopy or flow cytometry
BactoSpore™ stains are fluorescent dyes optimised for staining endospores. The dyes also stain both live and dead bacteria of gram-positive and gram-negative strains. In B. subtilis, the dyes stain both vegetative cells and endospores and have been validated for detection by fluorescence microscopy and flow cytometry.
Bacterial endospores are tough dormant structures formed by certain strains of bacteria including Bacillus and Clostridium species in response to nutrient deprivation and other stressors. Endospore formation allows these bacteria to survive in a non-replicative state until growth conditions improve, at which point the spores can germinate to allow vegetative cell replication.
Use BactoSpore™ 485/500 for membrane staining in the FITC channel.
Order Now
Specification Test Results
Ex/Em (nm) 488/536 (with DNA)
Detection Channel FITC or PE Channel