Procion® Red MX-5B, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Danger

Synonyms: Reactive red 2

404365-5G
404365-5GEA 51.4 EUR
404365-5G
Procion® Red MX-5B, Sigma-Aldrich®
Procion® Red MX-5B

Procion Red has been used in the visualization of osteocyte canalicular system. It has been used as an adsorbate in the biosorption by Bacillus subtilis.

Formula: C₁₉HCl₂N₆Na₂O₇S₂
MW: 615.33 g/mol
Melting Pt: >300 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00067094
CAS Number: 17804-49-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39d0a45d1ac7f290209718456784c3ca2b25d212&vid=020c7bf90096b578ae61770384bc69c22c3c1699&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR