Nickel 99,95 Metals basis, foil_thin, Thickness: 0.0025 mm (0.0001 in)

Supplier: Thermo Fisher Scientific
Danger

40714.FF 40714.FI 40714.GH
40714.FFEA 272 EUR
40714.FF 40714.FI 40714.GH
Nickel 99,95 Metals basis, foil_thin, Thickness: 0.0025 mm (0.0001 in)
Nickel

Purity: 99.95% CAS No.: 7440-02-0 Physical Form: ≈0.014g/25x25mm, not certified pinhole free Odour: Odourless MDL No.: MFCD00011137

Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR