Ammonium iron(II) sulphate hexahydrate 99,999 Metals basis, Puratronic®

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Ferrous Ammonium sulphate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate

40751.03 40751.06 40751.14
40751.03EA 42.5 EUR
40751.03 40751.06 40751.14
Ammonium iron(II) sulphate hexahydrate 99,999 Metals basis, Puratronic®
Ammonium iron (II) sulphate hexahydrate

Purity: 99.999% CAS No.: 7783-85-9 Molecular Formula: H8FeN2O8S2.xH2O (x≈6) Formula Weight: 284.04(anhy) Physical Form: Coarse Powder Density: 1.864 Appearance: Pale bluish green Odour: Odourless Melting Point: ca 100° dec. Merck Reference: 14,521 MDL No.: MFCD00150530

Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
MW: 392.14 g/mol
Melting Pt: ∼ 100 °C (dec.)
Density: 1.864
Storage Temperature: Ambient
MDL Number: MFCD00150530
CAS Number: 7783-85-9
EINECS: 233-382-4
Merck Index: 14,521

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR