Graphite 99,9995 Metals basis, rods, Length: 102mm (4in), Ø 6.15mm (0.242in)

Supplier: Thermo Fisher Scientific

40766.KD
40766.KDEA 347 EUR
40766.KD
Graphite 99,9995 Metals basis, rods, Length: 102mm (4in), Ø 6.15mm (0.242in)
Graphite
Formula: C
MW: 12.01 g/mol
Melting Pt: 3827
Density: 2.09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR