Grinding Jars for MM 200 and MM 400
Supplier: Retsch GmbH
01.462.0195 02.462.0059 02.462.0119 01.462.0008 01.462.0009 01.462.0113 01.462.0115 01.462.0194 02.462.0051 02.462.0052 02.462.0057 02.462.0061 01.462.0201 01.462.0213 01.462.0214 01.462.0215 01.462.0216 01.462.0217 01.462.0230 01.462.0231 01.462.0232 01.462.0233 01.462.0234 01.462.0235 01.462.0236 01.462.0237 01.462.0238 02.462.0184 01.462.0244
412-3004EA 1460 EUR
412-3004 412-3042 412-3044 412-3063 412-3053 412-3062 412-3052 412-0007 412-3074 412-3034 412-3041 412-3043 412-2119 412-2120 412-2127 412-2126 412-2128 412-2123 412-2112 412-2113 412-2114 412-2118 412-2115 412-2117 412-2116 412-2124 412-2125 412-0165 RETS01.462.0244
Grinding Jars for MM 200 and MM 400
Jars Grinding Jars/Bowls
A wide range of jars is available both in terms of size and choice of material.
  • The jars for the MM 200 have a push-fit lid, whereas the jars for the MM 400 have a screw cap lid
  • All grinding jars with screw top lids up to 10 ml and also the 25 ml stainless steel jar (412-0014) can be used with the MM 200
Upozornení: * Typically a jar filling should consist of 1/3 sample and 1/3 ball charge.