Manganese ≥99.6% (metals basis), powder -140+325 mesh

Supplier: Thermo Fisher Scientific
Warning

41343.14 41343.22 41343.36
41343.14EA 45.3 EUR
41343.14 41343.22 41343.36
Manganese ≥99.6% (metals basis), powder -140+325 mesh
Manganese

Purity: 99.6% CAS No.: 7439-96-5 UN No.: 3089 MDL No.: MFCD00011111

Formula: Mn
MW: 54.94 g/mol
Density: 7.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1
UN: 3089
ADR: 4.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR