Aluminium ≥99.0% (metals basis), thin foil, Thickness: 8.0 µm (0.0003 in)

Supplier: Thermo Fisher Scientific

Synonyms: Aluminum

41358.FIEA 93.1 EUR
41358.FI 41358.GH
Aluminium ≥99.0% (metals basis), thin foil, Thickness: 8.0 µm (0.0003 in)
Aluminium

Purity: 99.0% CAS No.: 7429-90-5 Physical Form: ≈0.054g/50x50mm, not certified pinhole free Odour: Odourless MDL No.: MFCD00134029

Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR