Silver ≥99.9% (metals basis), foil, annealed, Thickness: 0.28 mm (0.011 in)

Supplier: Thermo Fisher Scientific

41451.FIEA 82.8 EUR
41451.FI 41451.GH 41451.GW
Silver ≥99.9% (metals basis), foil, annealed, Thickness: 0.28 mm (0.011 in)
Silver

Purity: 99.9% CAS No.: 7440-22-4 Physical Form: ≈7.3g/50x50mm, Temper: soft Odour: Characteristic MDL No.: MFCD00003397

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR