Platinum rhodium alloy (90:10 w%) ≥99.9% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Supplier: Thermo Fisher Scientific
41810.FF 41810.FI 41810.GH
41810.FFEA 3120 EUR
41810.FF 41810.FI 41810.GH
Platinum rhodium alloy (90:10 w%) ≥99.9% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Platinum Rhodium alloy (90:10 w%)
Purity: 99.9% Molecular Formula: Pt:Rh; 90:10 wt% Physical Form: ≈1.25g/25x25mm Odour: Odourless MDL No.: MFCD00198186