VWR®, Bovine serum albumin (BSA)
Supplier: VWR Chemicals
422351SEA 616 EUR
422351S
VWR®, Bovine serum albumin (BSA)
Proteins and Peptides
A higher purity product than standard grade or Cohn fraction V material. It is negative for globulins that interfere in many assays.
  • Moisture: 4,5%
  • Ashes: 1,5%