Fused quartz microscope slides
Supplier: Thermo Fisher Scientific
42295.KG 42295.KJ 42296.KG 42296.KJ 42297.KG 42297.KJ
42295.KGEA 57.5 EUR
42295.KG 42295.KJ 42296.KG 42296.KJ 42297.KG 42297.KJ
Fused quartz microscope slides
Slides Microscope Slides
These fused quartz slides are made of fusing naturally occurring crystalline silica.
  • High chemical purity, high corrosion resistance, high melting point
  • Low coefficient of thermal expansion with excellent electrical insulation
  • Optical transmission from ultra-violet to infrared