Aluminium ≥99.99% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)

Supplier: Thermo Fisher Scientific

Synonyms: Aluminum

42327.09 42327.18 42327.30
42327.09EA 37.8 EUR
42327.09 42327.18 42327.30
Aluminium ≥99.99% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Aluminium

Purity: 99.99% CAS No.: 7429-90-5 Physical Form: ≈0.54g/slug Odour: Odourless MDL No.: MFCD00134029

Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR