Titanium ≥99.98% (metals basis), slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)

Supplier: Thermo Fisher Scientific
Warning

42390.09 42390.18
42390.09EA 108 EUR
42390.09 42390.18
Titanium ≥99.98% (metals basis), slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Titanium

Purity: 99.98% CAS No.: 7440-32-6 Physical Form: ≈0.11g/slug Odour: Odourless MDL No.: MFCD00011264


Evaporation source

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR