Barium standard dissolved in oil, 5,000 µg/g Ba in mineral oil for wear metal analysis, Specpure®

Supplier: Thermo Fisher Scientific

42434.18
42434.18EA 70 EUR
42434.18
Barium standard dissolved in oil, 5,000 µg/g Ba in mineral oil for wear metal analysis, Specpure®
Standards Cationic Standards Barium Standards

Physical Form: Liquid
MDL No.: MFCD00134031

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR