Graphite conductive adhesive, alcohol based

Supplier: Thermo Fisher Scientific
Danger

42465.15
42465.15EA 58.1 EUR
42465.15
Graphite conductive adhesive, alcohol based
Graphite conductive adhesive

Flash Point: 11°C(52°F) Physical Form: Sheet resistance: 1.2kohm/in2 @ 0.001in thick Density: 0.90 Appearance: Black Boiling Point: 82°C UN No.: UN1133 MDL No.: MFCD01866817

Boiling Pt: 82 °C
Density: 0.90
Flash Pt: 11 °C (52 °F)
Storage Temperature: Ambient
MDL Number: MFCD01866817
UN: 1133
ADR: 3 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR