Hydrogen chloride 1.25 M ethanol, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

427011000 427018000 427011000_T 427018000_T
427011000.EA 50.4 EUR
427011000. 427018000. 427011000_T 427018000_T
Hydrogen chloride 1.25 M ethanol, AcroSeal®
Hydrogen chloride
Formula: HCl
MW: 36.46 g/mol
Flash Pt: 12 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 2924
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Water ≤0.5 %
Strength 1.10 to 1.40 M
NMR ≥96.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR