Glassy carbon, plate, Thickness: 3 mm (0.1 in) Type 2

Supplier: Thermo Fisher Scientific

42820.FF 42820.FI
42820.FFEA 152 EUR
42820.FF 42820.FI
Glassy carbon, plate, Thickness: 3 mm (0.1 in) Type 2
Glassy carbon
Formula: C
MW: 12.01 g/mol
Melting Pt: 3652
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR