Sodium methoxide 30% w/w 5.4 M methanol, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Sodium methylate, Sodium methanolate

428361000 428368000
428361000.EA 35.8 EUR
428361000. 428368000.
Sodium methoxide 30% w/w 5.4 M methanol, AcroSeal®
Sodium methoxide
Formula: CH₃NaO
MW: 54.02 g/mol
Melting Pt: 1…5 °C
Density: 0.97 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00012179
CAS Number: 124-41-4
UN: 1289
ADR: 3 III

Order Now

Specification Test Results

Appearance Colorless to light yellow Solution
Strength 5.3 to 5.6 M
HPLC ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR