Magnesium chloride, dry_ultra 99,99 Metals basis

Supplier: Thermo Fisher Scientific
Warning


ampuled under argon

42850.06 42850.14 42850.22
42850.06EA 103 EUR
42850.06 42850.14 42850.22
Magnesium chloride, dry_ultra 99,99 Metals basis
Magnesium chloride

Purity: 99.99% CAS No.: 7786-30-3 Molecular Formula: Cl2Mg Formula Weight: 95.22 Physical Form: Powder, Ampouled under argon Density: 2.32 Appearance: White to yellow Odour: Odourless Melting Point: 714° Merck Reference: 14,5662 MDL No.: MFCD00011106

Formula: MgCl₂
MW: 95.21 g/mol
Boiling Pt: 1412
Melting Pt: 714 °C
Density: 2.32
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Merck Index: 14,5662

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR