Titanium 99,7 Metals basis, plate 100x100 mm (3.9x3.9 in), annealed, Thickness: 6.35 mm (0.25 in)

Supplier: Thermo Fisher Scientific
Warning

42896.KG 42896.KJ
42896.KGEA 193 EUR
42896.KG 42896.KJ
Titanium 99,7 Metals basis, plate 100x100 mm (3.9x3.9 in), annealed, Thickness: 6.35 mm (0.25 in)
Titanium

Purity: 99.7% CAS No.: 7440-32-6 Physical Form: ≈286g/pc Odour: Odourless MDL No.: MFCD00011264

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR