Bag openers for Stomacher® bags
Supplier: Seward (FermionX)
BA6095 BA6098
431-0132EA 49.7 EUR
431-0132 432-3115
Bag openers for Stomacher® bags
Racks Bag Racks
Made from high grade plastic polymer.
  • Compatible with Stomacher® 80 bags
  • Stable on bench-top or scales
  • Easily cleaned with most lab disinfectants
These bag openers hold open bags for easy addition of sample without risk of instrument contact on the bag.