Nickel 99,98 Metals basis, wire, annealed, Ø 1.0 mm (0.04 in)

Supplier: Thermo Fisher Scientific
Danger

43133.G6 43133.H2 43133.G2
43133.G6EA 190 EUR
43133.G6 43133.H2 43133.G2
Nickel 99,98 Metals basis, wire, annealed, Ø 1.0 mm (0.04 in)
Nickel

Purity: 99.98% CAS No.: 7440-02-0 Physical Form: ≈7.0g/m, Temper: soft Odour: Odourless MDL No.: MFCD00011137

Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR