Micro-tissue grinders with screw caps
Supplier: DWK Life Sciences
357535
432-5002EA 351 EUR
432-5002
Micro-tissue grinders with screw caps
Homogenisers Tissue Grinders
Borosilicate glass tube, PTFE pestle with stainless steel shaft.
13-425 GPI thread finish, cap has open top and PTFE-faced silicone rubber liner.
Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).
Clearance between pestle and mortar: 0,05 to 0,15 mm