Micro-tissue grinder set
Supplier: DWK Life Sciences
358204
432-5063EA 839 EUR
432-5063
Micro-tissue grinder set
Homogenisers Tissue Grinders
A complete selection of Wheaton Micro tissue grinders conveniently packed in a lightweight protective carrying case.
Includes the following tissue grinders:
- 1 ml Tenbroeck
- 0,5 ml grinder with 13 mm-425 GPI cap
- 1 ml Dounce
- 0,1 and 0,2 ml standard
- 2 ml Potter-Elvehjem
- 1 ml tapered