Polyvinylpyrrolidone, M.W. ∼ 1.300.000 (K85-95)

Supplier: Thermo Fisher Scientific

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

43728.A1 43728.30 43728.18
43728.A1EA 264 EUR
43728.A1 43728.30 43728.18
Polyvinylpyrrolidone, M.W. ∼ 1.300.000 (K85-95)
Polyvinylpyrrolidone

CAS No.: 9003-39-8 Molecular Formula: (C6H9NO)n Physical Form: Powder Sensitivity: Hygroscopic Odour: Odourless Merck Reference: 14,7697 MDL No.: MFCD00149016

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8
Merck Index: 14,7697

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR