VWR® VOS 40 digital & VOS 60 control, Electronic Overhead Stirrers
Supplier: VWR Collection
441-0304EA 1080 EUR
441-0304 441-0302 441-0303 441-0305 441-0306 441-0307
VWR® VOS 40 digital & VOS 60 control, Electronic Overhead Stirrers
Stirrers Overhead Stirrers
Laboratory stirrers that are ideal for simple stirring tasks, suitable for quantities up to 25 L (VOS 40 digital), or for intensive stirring tasks for quantities up to 40 L (VOS 60 control). Due to microprocessor-controlled technology, the stirrers automatically adjust the speed within the speed range of 0/30 to 2000 min⁻¹.
  • Digital speed display
  • Infinitely adjustable speed
  • Push-through agitator shafts
  • Overload protection - units can be operated for short periods in overload conditions
  • Error code display
Safety circuits ensure automaticMore Product Information
Order Now
SPECIFICATIONS
Model VOS 40 digital VOS 60 control
Max. stirring capacity H₂O 25 L 40 L
Environmentally Preferable
Max. viscosity 30000 mPas 50000 mPas
Speed range 0/30 - 2000 min⁻¹ 0/30 - 2000 min⁻¹
Max. torque at chuck 40 Ncm 60 Ncm
Chuck range 0,5 - 10 mm 0,5 - 10 mm
Power - motor input/output 112/87 W 168/131 W
W×D×H 86×208×248 mm 86×230×267 mm
Weight 4,4 kg 4,7 kg