Electronic overhead stirrers, Hei-TORQUE series
Supplier: Heidolph Instruments GmbH & Co.KG
501-62010-04 501-62020-00 501-62020-03 501-62020-04 501-64010-00 501-64010-03 501-64010-04 501-64020-00 501-64020-03 501-64020-04 501-61010-00 501-61010-03 501-61010-04 501-61020-00 501-61020-03 501-61020-04 501-62010-00 501-62010-03 501-62030-00 501-62020-00-2
441-0395EA 1740 EUR
441-0395 441-0396 441-0397 441-0398 441-0399 441-0400 441-0401 441-0402 441-0403 441-0404 441-0387 441-0388 441-0389 441-0390 441-0391 441-0392 441-0393 441-0394 HEID501-62030-00 HEID501-62020-00-2
Electronic overhead stirrers, Hei-TORQUE series
Stirrers Overhead Stirrers
Powerful electronic stirrers that feature a smooth start operation which prevents spills and splashing media. The speed ramps up slowly until set speed has been reached. Both models feature safe start and stop of operation via an intuitive glass slide touch panel to avoid accidental start-up. An overtemperature sensor shuts off the unit in dangerous heat-up situations, ideal for unattended continuous operation. All units are designed for continuous 24-hour operation, including challenging high viscosity applications in polymer research. High torque levels give better mixing results in lessMore Product Information