Tin ≥98.8% (metals basis), foil, Thickness: 0.1 mm (0.004 in)

Supplier: Thermo Fisher Scientific

44154.RG
44154.RGEA 491 EUR
44154.RG
Tin ≥98.8% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Tin

Purity: 98.8% CAS No.: 7440-31-5 Physical Form: ≈329g/30x150cm Odour: Odourless MDL No.: MFCD00133862

Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR