Magnetic stirrer, lab disc white
Supplier: IKA
3765000 3907500
IKAA3765000EA 0 EUR
IKAA3765000 442-0737
Magnetic stirrer, lab disc white
Stirrers Magnetic Stirrers
Ultra-flat magnetic stirrers with magnetic coil technology. Top plate and casing are made of chemically resistant materials. Wear-free drive with no moving parts.
  • Reverses direction of rotation every 30 seconds to ensure optimum mixing (selectable)
  • Can be used at 5 to 40 °C (80% relative humidity), in an incubator or cold room
IP protection class according to DIN EN 60529: IP 65
Certifikace: IP protection class according to DIN EN 60529: IP 65