Magnetic stirrers for cell cultures, bioMIXdrive series
Supplier: 2MAG
80002 80001 80003 80004 98400_UK 98604_UK 98604_CH 98604 98500_UK 98500 98400_CH 98400 98500_CH
442-0790EA 1228 EUR
442-0790 442-0789 442-0791 442-0792 442-0776 442-0782 442-0781 442-0780 442-0779 442-0777 442-0775 442-0774 442-0778
Magnetic stirrers for cell cultures, bioMIXdrive series
Stirrers Magnetic Stirrers, Multi-position
Stirring systems for careful mixing of cell cultures. Suitable for culture flasks with spinning propeller and spinning ball systems. The hermetically closed stainless steel housing is water, germ and dust proof.
  • Extremely slow with 100% jerk-free mixing
  • Submersible in water baths
  • Designed for long term operation in CO₂ incubators
IP protection class: IP 68.
External control unit (must be ordered separately).
SoftStart for reliable and safe acceleration of the stirring bar
QuickSet for setting of start and maximum speed
Storage of last workingMore Product Information