Multi-position magnetic stirrers, MIXdrive series
Supplier: 2MAG
40215 40206 40260 40060 40006 40015
442-0787EA 2312 EUR
442-0787 442-0786 442-0788 442-0785 442-0783 442-0784
Multi-position magnetic stirrers, MIXdrive series
Stirrers Magnetic Stirrers, Multi-position
The hermetically sealed stainless steel housing is water, germ and dust-proof. Operating temperatures for basic version up to 50 °C, for high temperature (HT) version up to 95 °C submerged in water baths, up to 200 °C in ovens.
  • Maintenance and wear-free inductive drive
  • 100% synchronised jerk-free mixing
  • Submersible in water and oil baths
  • Suitable for use in CO₂ incubator
SoftStart for reliable and safe acceleration of the stirring bar
QuickSet for setting of start and maximum speed
Storage of last working status when turning off
Digital display for speedMore Product Information