VWR®, Stirring Rods
Supplier: VWR Collection
442-4539EA 27.2 EUR
442-4539 442-4541 442-4542
VWR®, Stirring Rods
Stirrers Stirring Rods
Solid PTFE.
  • Inert and will not scratch, can be used up to 280 °C