Aluminium 99,9999 Metals basis, foil, Puratronic®, annealed, Thickness: 0.2 mm (0.008 in)

Supplier: Thermo Fisher Scientific

Synonyms: Aluminum

44330.GH 44330.HP
44330.GHEA 176 EUR
44330.GH 44330.HP
Aluminium 99,9999 Metals basis, foil, Puratronic®, annealed, Thickness: 0.2 mm (0.008 in)
Aluminium

Purity: 99.9999% CAS No.: 7429-90-5 Physical Form: ≈5.4g/100x100mm, Temper: soft Odour: Odourless MDL No.: MFCD00134029

Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR