Tube rotators
Supplier: Thermo Fisher Scientific
88881002
444-0983EA 475 EUR
444-0983
Tube rotators
Shakers Rotating/Rocking Shakers
This small footprint tube rotator is ideal for common applications such as molecular biology, clinical and biochemistry. The agitating mode achieves more effective mixing with oscillation occurring every 40 degrees in the rotation cycle.
  • Simple display
  • Interchangeable rotisseries accommodate different tube sizes
  • Each paddle can be adjusted from 0 to 90 degrees
  • Maintenance-free motor and plug-and-play simplicity
Certifikace: CE and cCSAus compliance.
Delivery information: Supplied with 2× 10/15, 2× 5/7, 2× 1,5/2 and 1× 50 ml paddles. ReplacementMore Product Information