Serological pipettes, Stripette™
Supplier: Corning
4011 4020 4021 4050 4100 4250 4484 4485 4486 4487 4488 4489 4490 4491 4492 4500 4501 7000 7015 7016 7017 7041 7042 7045 4101 4251 4051
734-1688EA 241.55 EUR
734-1688 734-1689 734-1690 734-1691 734-1693 734-1695 734-1734 734-1735 734-1736 734-1737 734-1738 734-1739 734-1740 734-1741 734-1742 734-1743 734-1744 444-1019 612-5123 612-5124 612-5125 612-5126 612-5127 612-5128 734-1694 734-1696 734-1692
Serological pipettes, Stripette™
Pipettes Serological Pipettes
PS, transparent, graduated, sterile.
  • Calibrated to deliver (TD, Ex)
  • High performance anti-drip tips, colour-coded magnifier stripe and negative graduations
  • Certified RNase-/DNase-free and non pyrogenic
* Wide orifice for viscous materials.
Paper/plastic wrap allows easy opening by either pop-through or peel-apart techniques. Ideal for sterile tissue culture applications. Wrap reduces static cling.
Exclusive anti-drip tip assures accurate delivery (25, 50, and 100 ml sizes). Colour-coded magnifier strips make volume reading easier. BidirectionalMore Product Information